Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other B3GALNT1 SIRNA Assays ›
Gene Symbol:
B3GALNT1
Gene Name:
beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
Gene Aliases:
B3GALT3, GLCT3, GLOB, Gb4Cer, P, P1, beta3Gal-T3, galT3
Chromosome Location:
Chr.3: 161083883 - 161105528 on Build GRCh38
UniGene ID:
Hs.418062
Species:
Human
siRNA ID
HSS112784
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: B3GALNT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001038628.1
NP_001033717.1
8
1227
NM_003781.3
NP_003772.1
5
904
NM_033167.2
NP_149357.1
6
1023
NM_033168.2
NP_149358.1
4
809
NM_033169.2
NP_149359.1
5
877
XM_005247859.4
XP_005247916.1
6
967
XM_005247861.3
XP_005247918.1
7
1087
XM_005247862.3
XP_005247919.1
6
945
XM_005247864.3
XP_005247921.1
5
902
XM_006713803.3
XP_006713866.1
7
1086
XM_006713804.3
XP_006713867.1
4
1027
XM_006713805.3
XP_006713868.1
5
1146
XM_017007402.1
XP_016862891.1
6
1041
XM_017007403.1
XP_016862892.1
6
989
XM_017007404.1
XP_016862893.1
6
997
XM_017007405.1
XP_016862894.1
5
870
XM_017007406.1
XP_016862895.1
7
1064
XM_017007407.1
XP_016862896.1
7
1213
XM_017007408.1
XP_016862897.1
6
1116
XM_017007409.1
XP_016862898.1
5
1009
XM_017007410.1
XP_016862899.1
7
1096
XM_017007411.1
XP_016862900.1
5
1018
XM_017007412.1
XP_016862901.1
4
943
GenBank mRNA
AB050855.1
-
1
566
AB050856.1
-
5
820
AB060691.1
-
1
235
AF132731.1
-
2
533
AF154848.1
-
5
853
AK022491.1
-
5
856
AK095917.1
-
4
809
AK223439.1
-
5
817
AY359049.1
-
4
779
BC028571.1
-
8
1149
4838965
BC047618.1
-
5
799
5259171
CR457170.1
-
1
458
Y15062.1
-
1
458
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
oligosaccharide biosynthetic process
UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
galactosylgalactosylglucosylceramide beta-D-acetylgalactosaminyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-vqgwm:80/10.239.167.106:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline