Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308007.1 | NP_001294936.1 | 8 | 1614 | ||
NM_003797.4 | NP_003788.2 | 8 | 1614 | |||
XM_011545330.1 | XP_011543632.1 | 8 | 1454 | |||
XM_011545331.1 | XP_011543633.1 | 8 | 1454 | |||
XM_017018512.1 | XP_016874001.1 | 8 | 1454 | |||
GenBank mRNA | AF070418.1 | - | 8 | 996 | ||
AF078933.1 | - | 8 | 760 | |||
AF080227.1 | - | 8 | 1112 | |||
AF099032.1 | - | 8 | 858 | |||
AK292120.1 | - | 8 | 1223 | |||
AK294133.1 | - | 8 | 661 | |||
AK301265.1 | - | 8 | 730 | |||
BC047672.2 | - | 8 | 727 | |||
BC068995.1 | - | 8 | 775 | 4637702 | ||
U90651.1 | - | 8 | 858 |