Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003825.3 | NP_003816.2 | 2 | 435 | ||
NM_130798.2 | NP_570710.1 | 2 | 435 | |||
XM_006720725.2 | XP_006720788.1 | 2 | 481 | |||
XM_017022692.1 | XP_016878181.1 | 2 | 481 | |||
XM_017022693.1 | XP_016878182.1 | 2 | 141 | |||
XM_017022694.1 | XP_016878183.1 | 3 | 203 | |||
GenBank mRNA | AJ011915.1 | - | 2 | 57 | ||
AK290152.1 | - | 2 | 481 | |||
AK313161.1 | - | 2 | 79 | |||
AK315279.1 | - | 2 | 435 | |||
BC000148.2 | - | 2 | 67 | 2900640 | ||
BC003686.1 | - | 2 | 86 | 3461227 | ||
BC022890.1 | - | 2 | 91 | 4658930 | ||
BI918294.1 | - | 2 | 103 | |||
BT006916.1 | - | 1 | 13 | |||
CR457212.1 | - | 1 | 13 | |||
DA650133.1 | - | 2 | 63 | |||
DA882401.1 | - | 2 | 435 | |||
U55936.1 | - | 2 | 105 | |||
Y09567.1 | - | 1 | 13 | |||
Y09568.1 | - | 1 | 13 |