Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003827.3 | NP_003818.2 | 5 | 717 | ||
NR_038456.1 | 5 | 717 | ||||
NR_038457.1 | 3 | 520 | ||||
XM_011527436.1 | XP_011525738.1 | 5 | 391 | |||
XM_011527437.1 | XP_011525739.1 | 4 | 470 | |||
GenBank mRNA | AK098401.1 | - | 5 | 550 | ||
AK126519.1 | - | 2 | 1604 | |||
AK292244.1 | - | 5 | 481 | |||
AK308633.1 | - | 6 | 607 | |||
AK310114.1 | - | 3 | 284 | |||
BC001165.1 | - | 5 | 489 | 3533668 | ||
BC007432.2 | - | 5 | 480 | 2962847 | ||
BC028234.1 | - | 5 | 535 | 5245770 | ||
BC091511.1 | - | 5 | 496 | 30512911 | ||
BT019568.1 | - | 5 | 399 | |||
CX873074.1 | - | 3 | 262 | |||
U39412.1 | - | 5 | 466 |