Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081955.2 | NP_001075424.1 | 12 | 1037 | ||
NM_001165933.1 | NP_001159405.1 | 12 | 1037 | |||
NM_003835.3 | NP_003826.2 | 12 | 1046 | |||
XM_011525426.2 | XP_011523728.1 | 5 | 1217 | |||
GenBank mRNA | AF071476.1 | - | 12 | 910 | ||
AF073710.1 | - | 3 | 771 | |||
AF493932.1 | - | 2 | 156 | |||
AF493933.1 | - | 12 | 843 | |||
AK289876.1 | - | 12 | 910 | |||
AK290535.1 | - | 12 | 906 | |||
AK301719.1 | - | 12 | 916 | |||
AK309061.1 | - | 12 | 927 | |||
AY585190.1 | - | 2 | 156 | |||
AY585191.1 | - | 12 | 834 | |||
BC096843.1 | - | 2 | 159 | 6519202 |