Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF10C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003841.3 | NP_003832.2 | 5 | 1049 | ||
GenBank mRNA | AF012536.1 | - | 5 | 933 | ||
AF012629.1 | - | 5 | 741 | |||
AF014794.1 | - | 5 | 890 | |||
AF016267.1 | - | 5 | 928 | |||
AF020502.1 | - | 5 | 861 | |||
AF033854.1 | - | 5 | 917 | |||
AK298664.1 | - | 4 | 836 | |||
AK298718.1 | - | 5 | 514 | |||
AK313213.1 | - | 5 | 943 | |||
AY358281.1 | - | 5 | 933 | |||
BC017852.1 | - | 5 | 942 | |||
BC125041.1 | - | 5 | 862 | 40115748 | ||
BC125042.1 | - | 5 | 862 | 40115749 | ||
CR541857.1 | - | 5 | 741 |