Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003889.3 | NP_003880.3 | 4 | 2305 | ||
NM_022002.2 | NP_071285.1 | 4 | 631 | |||
NM_033013.2 | NP_148934.1 | 4 | 2305 | |||
GenBank mRNA | AB307701.1 | - | 3 | 466 | ||
AF061056.1 | - | 4 | 769 | |||
AF084644.1 | - | 4 | 642 | |||
AF084645.1 | - | 4 | 745 | |||
AJ009936.1 | - | 5 | 2305 | |||
AJ009937.1 | - | 5 | 2305 | |||
AK122990.1 | - | 1 | 598 | |||
AK223456.1 | - | 4 | 750 | |||
AY091855.1 | - | 4 | 1048 | |||
BC017304.2 | - | 4 | 705 | 5087401 | ||
HQ692837.1 | - | 4 | 583 | |||
HQ709177.1 | - | 4 | 574 |