Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317086.1 | NP_001304015.1 | 12 | 1096 | ||
NM_003904.4 | NP_003895.1 | 13 | 1258 | |||
GenBank mRNA | AF019767.1 | - | 13 | 1238 | ||
AK301227.1 | - | 13 | 1206 | |||
BC004256.1 | - | 13 | 1213 | 3610932 | ||
BC012162.1 | - | 13 | 1226 | 4578136 | ||
BC017349.2 | - | 13 | 1243 | 5013820 | ||
BC017380.2 | - | 13 | 1226 | 2988688 | ||
BC111028.1 | - | 13 | 1230 | 5245870 | ||
BT006642.1 | - | 13 | 1210 |