Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111045.1 | NP_001104515.1 | 6 | 1273 | ||
NM_001111046.1 | NP_001104516.1 | 6 | 1155 | |||
NM_001111047.1 | NP_001104517.1 | 6 | 1152 | |||
NM_003914.3 | NP_003905.1 | 6 | 1276 | |||
XM_011535294.2 | XP_011533596.1 | 6 | 2107 | |||
XM_011535295.2 | XP_011533597.1 | 6 | 2104 | |||
XM_011535296.2 | XP_011533598.1 | 6 | 932 | |||
GenBank mRNA | AK301897.1 | - | 6 | 1048 | ||
AK314261.1 | - | 6 | 1276 | |||
AK316392.1 | - | 6 | 1075 | |||
BC035479.1 | - | 6 | 1022 | |||
BC036346.1 | - | 6 | 1022 | 5172478 | ||
BT019577.1 | - | 6 | 917 | |||
U66838.1 | - | 6 | 1055 | |||
U97680.1 | - | 5 | 765 |