Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111045.1 | NP_001104515.1 | 7, 8 | 1569 | ||
NM_001111046.1 | NP_001104516.1 | 7, 8 | 1451 | |||
NM_001111047.1 | NP_001104517.1 | 7, 8 | 1448 | |||
NM_003914.3 | NP_003905.1 | 7, 8 | 1572 | |||
XM_011535294.2 | XP_011533596.1 | 7, 8 | 2403 | |||
XM_011535295.2 | XP_011533597.1 | 7, 8 | 2400 | |||
XM_011535296.2 | XP_011533598.1 | 7, 8 | 1228 | |||
GenBank mRNA | AA553454.1 | - | 2 | 391 | ||
AK225146.1 | - | 5, 6 | 917 | |||
AK301897.1 | - | 7, 8 | 1344 | |||
AK314261.1 | - | 7, 8 | 1572 | |||
AK316392.1 | - | 7, 8 | 1371 | |||
BC035479.1 | - | 7, 8 | 1318 | |||
BC036346.1 | - | 7, 8 | 1318 | 5172478 | ||
BT019577.1 | - | 7, 8 | 1213 | |||
U66838.1 | - | 7, 8 | 1351 | |||
U97680.1 | - | 6, 7 | 1061 |