Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004667.5 | NP_004658.3 | 60 | 9294 | ||
XM_005268276.4 | XP_005268333.1 | 59 | 9424 | |||
XM_006720726.3 | XP_006720789.1 | 60 | 9300 | |||
XM_006720727.3 | XP_006720790.1 | 58 | 9057 | |||
XM_011522131.2 | XP_011520433.1 | 58 | 9081 | |||
XM_011522133.2 | XP_011520435.1 | 41 | 6060 | |||
XM_017022695.1 | XP_016878184.1 | 59 | 9241 | |||
XM_017022696.1 | XP_016878185.1 | 59 | 9184 | |||
XM_017022697.1 | XP_016878186.1 | 19 | 2623 | |||
XM_017022698.1 | XP_016878187.1 | 19 | 2534 | |||
GenBank mRNA | AF071172.3 | - | 60 | 9246 | ||
BC148318.2 | - | 14 | 1588 |