Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP7 SIRNA Assays ›
Gene Symbol:
MAP7
Gene Name:
microtubule associated protein 7
Gene Aliases:
E-MAP-115, EMAP115
Chromosome Location:
Chr.6: 136342281 - 136551015 on Build GRCh38
UniGene ID:
Hs.486548
Species:
Human
siRNA ID
HSS113376
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198608.1
NP_001185537.1
2
396
NM_001198609.1
NP_001185538.1
2
396
NM_001198611.1
NP_001185540.1
2
396
NM_001198614.1
NP_001185543.1
3
399
NM_001198615.1
NP_001185544.1
2
129
NM_001198616.1
NP_001185545.1
2
492
NM_001198617.1
NP_001185546.1
2
492
NM_001198618.1
NP_001185547.1
2
259
NM_001198619.1
NP_001185548.1
2
132
NM_003980.4
NP_003971.1
2
492
XM_006715598.2
XP_006715661.1
3
917
XM_006715599.2
XP_006715662.1
3
686
XM_006715600.2
XP_006715663.1
3
686
XM_006715601.3
XP_006715664.1
2
688
XM_011536243.2
XP_011534545.1
2
1287
XM_011536244.2
XP_011534546.1
2
1307
XM_011536245.1
XP_011534547.1
2
186
XM_011536246.2
XP_011534548.1
2
1288
XM_017011471.1
XP_016866960.1
3
917
GenBank mRNA
AJ242501.1
-
2
81
AJ242502.1
-
2
81
AK294461.1
-
2
166
AK295749.1
-
2
124
AK296483.1
-
2
259
AK296556.1
-
2
258
AK299355.1
-
2
129
AK307873.1
-
2
235
AK311227.1
-
2
411
AK311234.1
-
3
276
AK311236.1
-
3
234
AK311270.1
-
3
399
AK312789.1
-
2
92
AK316062.1
-
2
132
AK316529.1
-
3
265
AL042021.1
-
2
492
BC025777.1
-
2
88
5222987
CD557798.1
-
2
332
DA329576.1
-
2
303
DC325462.1
-
2
346
DC332469.1
-
2
411
X73882.1
-
2
181
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
response to osmotic stress
establishment or maintenance of cell polarity
protein localization to plasma membrane
receptor binding
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline