Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004685.3 | NP_004676.3 | 4, 5 | 781 | ||
XM_011535306.1 | XP_011533608.1 | 4, 5 | 727 | |||
XM_011535307.1 | XP_011533609.1 | 4, 5 | 727 | |||
XM_017020845.1 | XP_016876334.1 | 4, 5 | 727 | |||
GenBank mRNA | AF406619.1 | - | 4, 5 | 462 | ||
AK314587.1 | - | 4, 5 | 646 | |||
AL832017.2 | - | 4, 5 | 704 | |||
BC040012.1 | - | 4, 5 | 653 | 5495564 |