Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057749.2 | NP_477097.1 | 12 | 1264 | ||
XM_017013957.1 | XP_016869446.1 | 11 | 1589 | |||
XM_017013958.1 | XP_016869447.1 | 12 | 1406 | |||
XM_017013959.1 | XP_016869448.1 | 12 | 1508 | |||
GenBank mRNA | AF091433.1 | - | 11 | 1197 | ||
AF102778.1 | - | 10 | 1160 | |||
AF106690.1 | - | 10 | 1160 | |||
AF112857.1 | - | 10 | 1025 | |||
BC007015.1 | - | 10 | 999 | 3997513 | ||
BC020729.1 | - | 12 | 1235 | 4774554 |
Gene Symbol: INTS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017013618.1 | XP_016869107.1 | 24 | 4132 | ||
XM_017013619.1 | XP_016869108.1 | 27 | 4538 |