Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003970.3 | NP_003961.3 | 22, 23 | 3033 | ||
XM_006716237.1 | XP_006716300.1 | 22, 23 | 2998 | |||
GenBank mRNA | AK297890.1 | - | 9, 10 | 1351 | ||
BC052969.1 | - | 22, 23 | 2986 | 6205359 | ||
BP258208.1 | - | 3, 4 | 385 | |||
BX501741.1 | - | 2, 3 | 222 | |||
DA900083.1 | - | 1, 2 | 23 | |||
X69089.1 | - | 22, 23 | 2943 |