Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135669.1 | NP_001129141.1 | 8, 9 | 1154 | ||
NM_004736.3 | NP_004727.2 | 8, 9 | 1154 | |||
GenBank mRNA | AF089744.1 | - | 8, 9 | 1127 | ||
AF099082.1 | - | 8, 9 | 1131 | |||
AF115389.1 | - | 8, 9 | 962 | |||
AK309750.1 | - | 8, 9 | 1142 | |||
AL133058.1 | - | 5, 6 | 656 | |||
AL137583.1 | - | 1, 2 | 127 | |||
BC041142.1 | - | 8, 9 | 1132 | 5582839 |
Set Membership: |
![]() |