Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135669.1 | NP_001129141.1 | 9 | 1186 | ||
NM_004736.3 | NP_004727.2 | 9 | 1186 | |||
GenBank mRNA | AF089744.1 | - | 9 | 1159 | ||
AF099082.1 | - | 9 | 1163 | |||
AF115389.1 | - | 9 | 994 | |||
AK309750.1 | - | 9 | 1174 | |||
AL133058.1 | - | 6 | 688 | |||
AL137583.1 | - | 2 | 159 | |||
BC041142.1 | - | 9 | 1164 | 5582839 |
Set Membership: |
![]() |