Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135669.1 | NP_001129141.1 | 7, 8 | 952 | ||
NM_004736.3 | NP_004727.2 | 7, 8 | 952 | |||
GenBank mRNA | AF089744.1 | - | 7, 8 | 925 | ||
AF099082.1 | - | 7, 8 | 929 | |||
AF115389.1 | - | 7, 8 | 760 | |||
AK309750.1 | - | 7, 8 | 940 | |||
AL133058.1 | - | 4, 5 | 454 | |||
BC041142.1 | - | 7, 8 | 930 | 5582839 |
Set Membership: |
![]() |