Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005450.4 | NP_005441.1 | 1 | 1202 | ||
GenBank mRNA | AK314080.1 | - | 1 | 1202 | ||
BC034027.1 | - | 1 | 987 | 4737725 |