Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004226.3 | NP_004217.1 | 5 | 894 | ||
XM_011512168.2 | XP_011510470.1 | 5 | 859 | |||
XM_011512169.1 | XP_011510471.1 | 5 | 892 | |||
XM_011512170.1 | XP_011510472.1 | 6 | 875 | |||
XM_011512171.2 | XP_011510473.1 | 4 | 387 | |||
GenBank mRNA | AB011421.1 | - | 5 | 831 | ||
AK312865.1 | - | 5 | 892 | |||
BC016040.1 | - | 5 | 833 | 4575012 | ||
CR542210.1 | - | 4 | 570 | |||
CR542225.1 | - | 4 | 570 |