Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018714.2 | NP_061184.1 | 11 | 2578 | ||
GenBank mRNA | AB037802.1 | - | 11 | 2540 | ||
AK025633.1 | - | 8 | 1704 | |||
AK098224.1 | - | 11 | 2298 | |||
AL359611.1 | - | 5 | 918 | |||
BC021985.2 | - | 11 | 2543 | 2821581 | ||
BC047465.1 | - | 12 | 2880 | 5285077 |