Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004273.4 | NP_004264.2 | 2 | 495 | ||
XM_006718075.3 | XP_006718138.1 | 2 | 728 | |||
XM_011540369.2 | XP_011538671.1 | 2 | 2421 | |||
GenBank mRNA | AB012192.1 | - | 2 | 489 | ||
AB017915.1 | - | 2 | 495 | |||
BC093690.1 | - | 1 | 104 | 7939535 | ||
BC104856.1 | - | 1 | 104 | 8143859 |