Search Thermo Fisher Scientific
Gene Symbol: ADAMTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320336.1 | NP_001307265.1 | 4, 5 | 1692 | ||
NM_005099.5 | NP_005090.3 | 4, 5 | 1692 | |||
GenBank mRNA | AB014588.1 | - | 4, 5 | 1666 | ||
AF148213.1 | - | 4, 5 | 1672 | |||
AK291573.1 | - | 4, 5 | 1691 | |||
AY358886.1 | - | 4, 5 | 1684 | |||
BC063293.1 | - | 4, 5 | 1691 | 5303796 |