Gene Symbol: ADAMTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014243.2 | NP_055058.2 | 3, 4 | 535 | ||
XM_011532421.2 | XP_011530723.1 | 3, 4 | 567 | |||
XM_011532422.2 | XP_011530724.1 | 3, 4 | 1066 | |||
GenBank mRNA | AB002364.1 | - | 3, 4 | 487 | ||
AF247668.1 | - | 3, 4 | 535 | |||
BC130287.1 | - | 3, 4 | 532 | 40146318 | ||
BC132735.1 | - | 3, 4 | 532 | 40146757 |