Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PMPCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004279.2 | NP_004270.2 | 10, 11 | 1274 | ||
XM_005250717.3 | XP_005250774.1 | 10, 11 | 1288 | |||
XM_006716181.3 | XP_006716244.1 | 10, 11 | 1287 | |||
GenBank mRNA | AF054182.1 | - | 10, 11 | 1253 | ||
AK000994.1 | - | 10, 11 | 1256 | |||
AK057656.1 | - | 9, 10 | 1072 | |||
AK098616.1 | - | 10, 11 | 1244 | |||
AK289864.1 | - | 10, 11 | 1256 | |||
AK297354.1 | - | 10, 11 | 1639 | |||
AL050216.1 | - | 6, 7 | 723 | |||
BC014079.2 | - | 10, 11 | 1214 | |||
BQ021474.1 | - | 3, 4 | 535 |