Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077261.3 | NP_001070729.2 | 45 | 7216 | ||
NM_001206654.1 | NP_001193583.1 | 45 | 7216 | |||
NM_006312.5 | NP_006303.4 | 46 | 7246 | |||
GenBank mRNA | AF113003.1 | - | 45 | 6935 | ||
AF125672.1 | - | 44 | 7060 | |||
AY965853.1 | - | 44 | 7060 | |||
BC004326.1 | - | 7 | 1201 | |||
S83390.1 | - | 12 | 2283 |