Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTRF1 SIRNA Assays ›
Gene Symbol:
MTRF1
Gene Name:
mitochondrial translational release factor 1
Gene Aliases:
MRF1, MTTRF1, RF1
Chromosome Location:
Chr.13: 41216369 - 41311794 on Build GRCh38
UniGene ID:
Hs.382176
Species:
Human
siRNA ID
HSS114362
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTRF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004294.2
NP_004285.2
9
1307
XM_005266599.2
XP_005266656.1
11
1556
XM_006719897.2
XP_006719960.1
11
1664
XM_006719898.2
XP_006719961.1
11
1648
XM_006719900.2
XP_006719963.1
10
1377
XM_011535315.2
XP_011533617.1
11
1773
XM_011535317.2
XP_011533619.1
10
1656
XM_011535318.2
XP_011533620.1
11
1816
XM_011535320.2
XP_011533622.1
11
1752
XM_011535321.2
XP_011533623.1
14
1888
XM_011535323.2
XP_011533625.1
11
1618
XM_011535325.2
XP_011533627.1
10
1571
XM_017020856.1
XP_016876345.1
10
1617
XM_017020857.1
XP_016876346.1
13
1751
XM_017020858.1
XP_016876347.1
12
1746
XM_017020859.1
XP_016876348.1
10
1617
XM_017020860.1
XP_016876349.1
12
1634
XM_017020861.1
XP_016876350.1
12
1671
XM_017020862.1
XP_016876351.1
11
2829
XM_017020863.1
XP_016876352.1
11
2677
XM_017020864.1
XP_016876353.1
11
2569
XM_017020865.1
XP_016876354.1
10
2521
XM_017020866.1
XP_016876355.1
9
2386
XM_017020867.1
XP_016876356.1
11
1534
XM_017020868.1
XP_016876357.1
9
1502
XM_017020869.1
XP_016876358.1
10
1400
XM_017020870.1
XP_016876359.1
11
1664
XM_017020871.1
XP_016876360.1
10
1377
XM_017020873.1
XP_016876362.1
14
1958
XM_017020874.1
XP_016876363.1
11
1510
XM_017020875.1
XP_016876364.1
11
1504
XM_017020876.1
XP_016876365.1
10
2844
XM_017020877.1
XP_016876366.1
12
1679
GenBank mRNA
AF072934.1
-
9
1278
AK055086.1
-
11
1678
AK294343.1
-
10
1464
BC042196.1
-
9
1307
5198308
Back To Top
More Information
Panther Classification:
Molecular Function -
translation release factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translational termination
mitochondrial translational termination
translation release factor activity
translation release factor activity, codon specific
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0