Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS2ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012262.3 | NP_036394.1 | 7 | 1331 | ||
GenBank mRNA | AB007917.1 | - | 7 | 1228 | ||
AB024568.1 | - | 7 | 1300 | |||
AK023260.1 | - | 7 | 1304 |