Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014648.3 | NP_055463.1 | 28 | 3517 | ||
XM_005247914.3 | XP_005247971.1 | 28 | 3316 | |||
XM_005247915.3 | XP_005247972.1 | 28 | 3286 | |||
XM_005247916.3 | XP_005247973.1 | 29 | 3458 | |||
XM_005247917.3 | XP_005247974.1 | 28 | 3247 | |||
XM_005247918.3 | XP_005247975.1 | 28 | 3734 | |||
XM_005247919.3 | XP_005247976.1 | 27 | 2868 | |||
XM_017007540.1 | XP_016863029.1 | 28 | 3363 | |||
GenBank mRNA | AB014575.1 | - | 28 | 3224 | ||
AF279370.1 | - | 13 | 1176 | |||
AK023138.1 | - | 28 | 3285 | |||
AK294967.1 | - | 17 | 1580 | |||
AK303942.1 | - | 15 | 1670 | |||
AY227652.1 | - | 15 | 1428 | |||
AY495710.1 | - | 3 | 298 | |||
BC063882.1 | - | 28 | 3270 | 30387514 |