Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KIAA0040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162893.1 | NP_001156365.1 | 5 | 1172 | ||
NM_001162894.1 | NP_001156366.1 | 4 | 1095 | |||
NM_001162895.1 | NP_001156367.1 | 3 | 961 | |||
NM_001319230.1 | NP_001306159.1 | 3 | 1021 | |||
NM_001319231.1 | NP_001306160.1 | 5 | 1110 | |||
NM_014656.2 | NP_055471.2 | 4 | 1035 | |||
XM_017002909.1 | XP_016858398.1 | 4 | 1522 | |||
XM_017002910.1 | XP_016858399.1 | 4 | 1302 | |||
XM_017002911.1 | XP_016858400.1 | 4 | 1305 | |||
GenBank mRNA | AK292691.1 | - | 4 | 1023 | ||
AL049799.1 | - | 4 | 1015 | |||
AL049800.1 | - | 4 | 945 | |||
BC032361.1 | - | 4 | 953 | |||
BC111699.1 | - | 3 | 876 | |||
D25539.1 | - | 4 | 910 |