Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162893.1 | NP_001156365.1 | 5 | 1198 | ||
NM_001162894.1 | NP_001156366.1 | 4 | 1121 | |||
NM_001162895.1 | NP_001156367.1 | 3 | 987 | |||
NM_001319230.1 | NP_001306159.1 | 3 | 1047 | |||
NM_001319231.1 | NP_001306160.1 | 5 | 1136 | |||
NM_014656.2 | NP_055471.2 | 4 | 1061 | |||
XM_017002909.1 | XP_016858398.1 | 4 | 1548 | |||
XM_017002910.1 | XP_016858399.1 | 4 | 1328 | |||
XM_017002911.1 | XP_016858400.1 | 4 | 1331 | |||
GenBank mRNA | AK292691.1 | - | 4 | 1049 | ||
AL049799.1 | - | 4 | 1041 | |||
AL049800.1 | - | 4 | 971 | |||
BC032361.1 | - | 4 | 979 | |||
BC111699.1 | - | 3 | 902 | |||
D25539.1 | - | 4 | 936 |