Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPIP5K1 SIRNA Assays ›
Gene Symbol:
PPIP5K1
Gene Name:
diphosphoinositol pentakisphosphate kinase 1
Gene Aliases:
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
Chromosome Location:
Chr.15: 43533462 - 43590272 on Build GRCh38
UniGene ID:
Hs.156814
Species:
Human
siRNA ID
HSS114489
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPIP5K1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130858.2
NP_001124330.1
31
4245
NM_001130859.2
NP_001124331.1
29
4165
NM_001190214.1
NP_001177143.1
30
4164
NM_014659.5
NP_055474.3
29
4088
XM_005254803.1
XP_005254860.1
31
4233
XM_005254804.1
XP_005254861.1
30
4170
XM_006720786.2
XP_006720849.1
29
4044
XM_011522249.1
XP_011520551.1
32
4383
XM_011522250.1
XP_011520552.1
32
4486
XM_011522252.1
XP_011520554.1
31
4357
XM_011522254.1
XP_011520556.1
31
4253
XM_011522258.1
XP_011520560.1
26
3643
XM_011522259.1
XP_011520561.1
31
4209
XM_017022735.1
XP_016878224.1
31
4700
XM_017022736.1
XP_016878225.1
31
4668
XM_017022737.1
XP_016878226.1
31
4349
XM_017022738.1
XP_016878227.1
31
4273
XM_017022739.1
XP_016878228.1
30
4234
XM_017022740.1
XP_016878229.1
31
4336
XM_017022741.1
XP_016878230.1
30
4549
XM_017022742.1
XP_016878231.1
31
4341
XM_017022743.1
XP_016878232.1
30
4207
XM_017022744.1
XP_016878233.1
30
4148
XM_017022745.1
XP_016878234.1
31
4312
XM_017022746.1
XP_016878235.1
30
4183
XM_017022747.1
XP_016878236.1
29
4108
XM_017022748.1
XP_016878237.1
29
4486
XM_017022749.1
XP_016878238.1
30
4246
XM_017022750.1
XP_016878239.1
29
4087
XM_017022751.1
XP_016878240.1
29
4040
XM_017022752.1
XP_016878241.1
30
4146
XM_017022753.1
XP_016878242.1
29
4120
XM_017022754.1
XP_016878243.1
29
4152
XM_017022755.1
XP_016878244.1
28
4018
XM_017022756.1
XP_016878245.1
28
3956
XM_017022757.1
XP_016878246.1
28
3914
XM_017022758.1
XP_016878247.1
24
3430
XM_017022760.1
XP_016878249.1
30
5681
GenBank mRNA
AB002375.1
-
29
4086
AF502586.1
-
30
4126
AF502587.1
-
29
4092
AF502588.1
-
29
4060
AF502589.1
-
28
3450
AF543190.1
-
28
3400
BC035611.1
-
9
1473
BC057395.1
-
30
4191
6142663
BX538022.1
-
29
4116
BX647814.1
-
30
4122
BX647870.1
-
8
1341
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
inositol metabolic process
phosphorylation
dephosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
acid phosphatase activity
ATP binding
diphosphoinositol-pentakisphosphate kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-dvnl5:80/10.239.162.83:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0