Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195555.1 | NP_001182484.1 | 12 | 2009 | ||
NM_001195556.1 | NP_001182485.1 | 12 | 1986 | |||
NM_014666.3 | NP_055481.1 | 12 | 1955 | |||
XM_011534701.2 | XP_011533003.1 | 12 | 2020 | |||
XM_017010087.1 | XP_016865576.1 | 12 | 1966 | |||
GenBank mRNA | AF434813.1 | - | 12 | 1750 | ||
AK092765.1 | - | 12 | 1956 | |||
AK128594.1 | - | 2 | 2236 | |||
AK300257.1 | - | 12 | 1977 | |||
AL833548.1 | - | 2 | 1418 | |||
BC004467.2 | - | 12 | 1816 | 3831533 | ||
BC013091.1 | - | 5 | 820 | |||
BC030264.1 | - | 4 | 1911 | |||
D79993.1 | - | 12 | 1851 | |||
KU178486.1 | - | 12 | 1750 | |||
KU178487.1 | - | 12 | 1804 |