Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS2 SIRNA Assays ›
Gene Symbol:
RIMS2
Gene Name:
regulating synaptic membrane exocytosis 2
Gene Aliases:
OBOE, RAB3IP3, RIM2
Chromosome Location:
Chr.8: 103500026 - 104254430 on Build GRCh38
UniGene ID:
Hs.655271
Species:
Human
siRNA ID
HSS114539
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100117.2
NP_001093587.1
12
2476
NM_001282881.1
NP_001269810.1
9
2174
NM_014677.4
NP_055492.3
9
2033
XM_005251106.3
XP_005251163.1
11
2569
XM_005251107.3
XP_005251164.1
11
2569
XM_011517395.2
XP_011515697.1
16
2670
XM_011517398.2
XP_011515700.1
16
2670
XM_017014006.1
XP_016869495.1
15
2622
XM_017014007.1
XP_016869496.1
16
2670
XM_017014008.1
XP_016869497.1
16
2670
XM_017014009.1
XP_016869498.1
16
2670
XM_017014010.1
XP_016869499.1
16
2529
XM_017014011.1
XP_016869500.1
15
2622
XM_017014012.1
XP_016869501.1
15
2622
XM_017014013.1
XP_016869502.1
14
2580
XM_017014014.1
XP_016869503.1
11
2428
XM_017014015.1
XP_016869504.1
16
2529
XM_017014016.1
XP_016869505.1
15
2481
XM_017014017.1
XP_016869506.1
15
2481
XM_017014018.1
XP_016869507.1
11
2428
XM_017014019.1
XP_016869508.1
13
2560
XM_017014020.1
XP_016869509.1
12
2995
XM_017014021.1
XP_016869510.1
15
2481
XM_017014022.1
XP_016869511.1
9
2093
XM_017014023.1
XP_016869512.1
15
2622
XM_017014024.1
XP_016869513.1
16
2670
XM_017014025.1
XP_016869514.1
15
2622
XM_017014026.1
XP_016869515.1
13
2842
XM_017014027.1
XP_016869516.1
15
2481
XM_017014028.1
XP_016869517.1
16
2529
XM_017014029.1
XP_016869518.1
12
3303
XM_017014030.1
XP_016869519.1
11
2569
XM_017014031.1
XP_016869520.1
16
2670
XM_017014032.1
XP_016869521.1
15
2481
XM_017014033.1
XP_016869522.1
14
3281
XM_017014034.1
XP_016869523.1
13
3210
XM_017014035.1
XP_016869524.1
12
3161
XM_017014036.1
XP_016869525.1
9
1952
XM_017014037.1
XP_016869526.1
12
3161
XM_017014038.1
XP_016869527.1
11
2428
XM_017014039.1
XP_016869528.1
15
3330
XM_017014040.1
XP_016869529.1
9
1952
XM_017014041.1
XP_016869530.1
9
1952
XM_017014042.1
XP_016869531.1
11
2428
XM_017014043.1
XP_016869532.1
9
2093
XM_017014044.1
XP_016869533.1
10
2000
XM_017014045.1
XP_016869534.1
9
1952
XM_017014046.1
XP_016869535.1
9
1952
GenBank mRNA
AB018294.1
-
9
2102
AF007156.1
-
6
518
AK126939.1
-
12
2476
AK307790.1
-
9
2000
AK308831.1
-
9
1833
BC043144.2
-
9
1949
5288275
CR749374.1
-
10
1914
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
positive regulation of gene expression
calcium ion regulated exocytosis
regulation of exocytosis
cAMP-mediated signaling
insulin secretion
regulation of membrane potential
calcium ion-regulated exocytosis of neurotransmitter
spontaneous neurotransmitter secretion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
Rab GTPase binding
ion channel binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0