Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST18 SIRNA Assays ›
Gene Symbol:
ST18
Gene Name:
ST18, C2H2C-type zinc finger
Gene Aliases:
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location:
Chr.8: 52110832 - 52409879 on Build GRCh38
UniGene ID:
Hs.655499
Species:
Human
siRNA ID
HSS114555
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014682.2
NP_055497.1
24
3632
XM_006716487.1
XP_006716550.1
24
3399
XM_011517629.1
XP_011515931.1
25
3555
XM_011517631.1
XP_011515933.1
26
3596
XM_011517632.1
XP_011515934.1
24
3410
XM_011517633.1
XP_011515935.1
24
3403
XM_011517634.1
XP_011515936.1
24
3599
XM_011517635.1
XP_011515937.1
24
3603
XM_011517636.2
XP_011515938.1
23
3680
XM_011517637.1
XP_011515939.1
22
3137
XM_011517638.2
XP_011515940.1
21
3178
XM_011517641.1
XP_011515943.1
26
3643
XM_011517642.1
XP_011515944.1
24
3309
XM_017014047.1
XP_016869536.1
24
3759
XM_017014048.1
XP_016869537.1
24
3763
XM_017014049.1
XP_016869538.1
25
3809
XM_017014050.1
XP_016869539.1
24
3596
XM_017014051.1
XP_016869540.1
23
3603
XM_017014052.1
XP_016869541.1
25
3813
XM_017014053.1
XP_016869542.1
25
3800
XM_017014054.1
XP_016869543.1
23
3849
XM_017014055.1
XP_016869544.1
23
3440
XM_017014056.1
XP_016869545.1
26
3850
XM_017014057.1
XP_016869546.1
24
3653
XM_017014058.1
XP_016869547.1
24
3414
XM_017014059.1
XP_016869548.1
24
3600
XM_017014060.1
XP_016869549.1
25
3637
XM_017014061.1
XP_016869550.1
23
3772
XM_017014062.1
XP_016869551.1
22
3524
XM_017014063.1
XP_016869552.1
23
4070
XM_017014064.1
XP_016869553.1
23
3684
XM_017014065.1
XP_016869554.1
25
3804
XM_017014066.1
XP_016869555.1
23
3443
XM_017014067.1
XP_016869556.1
23
3853
XM_017014068.1
XP_016869557.1
24
3644
XM_017014069.1
XP_016869558.1
21
3341
XM_017014070.1
XP_016869559.1
20
3262
XM_017014071.1
XP_016869560.1
21
3510
XM_017014072.1
XP_016869561.1
25
3674
XM_017014073.1
XP_016869562.1
25
3312
XM_017014074.1
XP_016869563.1
24
3595
XM_017014075.1
XP_016869564.1
25
3846
XM_017014076.1
XP_016869565.1
25
3850
XM_017014077.1
XP_016869566.1
25
3487
XM_017014078.1
XP_016869567.1
24
3767
XM_017014079.1
XP_016869568.1
24
4158
XM_017014080.1
XP_016869569.1
22
3399
XM_017014081.1
XP_016869570.1
23
3520
XM_017014082.1
XP_016869571.1
23
3355
XM_017014083.1
XP_016869572.1
22
3195
XM_017014084.1
XP_016869573.1
20
2933
GenBank mRNA
AB011107.1
-
24
3496
BC117147.1
-
19
2889
40125698
BC117149.1
-
19
2889
40125700
BC143872.1
-
19
2884
BC143873.1
-
20
2977
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0