Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010989.2 | NP_001010989.1 | 6 | 1019 | ||
NM_014685.3 | NP_055500.1 | 6 | 1022 | |||
GenBank mRNA | AB034989.1 | - | 6 | 965 | ||
AF055001.1 | - | 6 | 962 | |||
AF055003.1 | - | 6 | 762 | |||
AF217990.1 | - | 3 | 1303 | |||
AK223233.1 | - | 6 | 965 | |||
AK291565.1 | - | 6 | 965 | |||
AK292088.1 | - | 6 | 965 | |||
AK302167.1 | - | 5 | 926 | |||
BC000086.1 | - | 6 | 957 | 3504515 | ||
BC008320.1 | - | 6 | 956 | 3350209 | ||
BC032673.1 | - | 6 | 964 | 5575914 | ||
CR457116.1 | - | 6 | 867 | |||
D14695.1 | - | 6 | 960 |