Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178129.1 | NP_001171600.1 | 14 | 1613 | ||
NM_012431.2 | NP_036563.1 | 14 | 2249 | |||
GenBank mRNA | AB002329.1 | - | 14 | 2094 | ||
AK297209.1 | - | 14 | 1929 | |||
AK303925.1 | - | 14 | 1614 | |||
BC140706.1 | - | 14 | 1784 | 9021574 | ||
BC144338.1 | - | 14 | 1784 | 9052858 | ||
BC152458.1 | - | 14 | 2106 | 8862593 |