Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG13 SIRNA Assays ›
Gene Symbol:
ATG13
Gene Name:
autophagy related 13
Gene Aliases:
KIAA0652, PARATARG8
Chromosome Location:
Chr.11: 46617276 - 46676019 on Build GRCh38
UniGene ID:
Hs.127403
Species:
Human
siRNA ID
HSS114714
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142673.2
NP_001136145.1
8, 9
1189
NM_001205119.1
NP_001192048.1
7, 8
1133
NM_001205120.1
NP_001192049.1
7, 8
1133
NM_001205121.1
NP_001192050.1
8, 9
1184
NM_001205122.1
NP_001192051.1
6, 7
1051
NM_014741.4
NP_055556.2
8, 9
1189
XM_005253262.2
XP_005253319.1
8, 9
929
XM_005253263.2
XP_005253320.1
8, 9
1220
XM_005253265.2
XP_005253322.1
9, 10
1426
XM_005253266.2
XP_005253323.1
8, 9
1155
XM_005253268.2
XP_005253325.1
6, 7
532
XM_006718394.2
XP_006718457.1
9, 10
1276
XM_006718395.2
XP_006718458.1
9, 10
1211
XM_006718396.2
XP_006718459.1
8, 9
1310
XM_011520493.1
XP_011518795.1
10, 11
1413
XM_011520496.2
XP_011518798.1
7, 8
1188
XM_011520499.2
XP_011518801.1
7, 8
802
XM_011520500.1
XP_011518802.1
9, 10
1276
XM_011520501.1
XP_011518803.1
9, 10
1276
XM_011520502.1
XP_011518804.1
9, 10
1276
XM_011520503.1
XP_011518805.1
9, 10
1276
XM_017018599.1
XP_016874088.1
9, 10
1361
XM_017018600.1
XP_016874089.1
8, 9
929
XM_017018601.1
XP_016874090.1
7, 8
1104
XM_017018602.1
XP_016874091.1
9, 10
1426
XM_017018603.1
XP_016874092.1
10, 11
1543
XM_017018604.1
XP_016874093.1
8, 9
1220
XM_017018605.1
XP_016874094.1
9, 10
1211
XM_017018606.1
XP_016874095.1
8, 9
1155
XM_017018607.1
XP_016874096.1
6, 7
532
XM_017018608.1
XP_016874097.1
8, 9
929
XM_017018609.1
XP_016874098.1
8, 9
1220
XM_017018610.1
XP_016874099.1
7, 8
1104
XM_017018611.1
XP_016874100.1
8, 9
1155
XM_017018612.1
XP_016874101.1
9, 10
1426
XM_017018613.1
XP_016874102.1
8, 9
1220
XM_017018614.1
XP_016874103.1
9, 10
1211
XM_017018615.1
XP_016874104.1
8, 9
1310
XM_017018616.1
XP_016874105.1
7, 8
1104
XM_017018617.1
XP_016874106.1
9, 10
1309
GenBank mRNA
AB014552.1
-
7, 8
813
AK023867.1
-
8, 9
922
AK223603.1
-
7, 8
809
AK289641.1
-
8, 9
736
AK293109.1
-
8, 9
888
AK294110.1
-
6, 7
764
BC001331.1
-
8, 9
618
3458359
BC002378.2
-
8, 9
888
2961127
BC006191.2
-
8, 9
930
3936851
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
macroautophagy
mitophagy in response to mitochondrial depolarization
positive regulation of protein targeting to mitochondrion
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0