Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135773.1 | NP_001129245.1 | 3 | 1805 | ||
NM_006038.3 | NP_006029.1 | 3 | 1721 | |||
XM_006723894.1 | XP_006723957.1 | 3 | 1641 | |||
XM_011529116.1 | XP_011527418.1 | 3 | 1678 | |||
GenBank mRNA | AB018300.1 | - | 3 | 1639 | ||
AK023229.1 | - | 3 | 1676 | |||
AK289644.1 | - | 3 | 1805 | |||
AK295536.1 | - | 2 | 1283 | |||
BC009481.2 | - | 3 | 1628 | 3953522 | ||
U28164.1 | - | 3 | 1686 |