Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256864.1 | NP_001243793.1 | 7 | 1067 | ||
NM_001256865.1 | NP_001243794.1 | 8 | 1015 | |||
NM_014787.3 | NP_055602.1 | 7 | 902 | |||
GenBank mRNA | AB007942.1 | - | 7 | 842 | ||
AK296408.1 | - | 8 | 969 | |||
BC051722.1 | - | 7 | 851 | |||
BC109279.2 | - | 7 | 1000 | 40028812 | ||
BC109280.1 | - | 7 | 1003 | 40028814 | ||
BX647203.1 | - | 7 | 1056 |