Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014838.2 | NP_055653.2 | 2 | 2954 | ||
GenBank mRNA | AB014537.1 | - | 2 | 2922 | ||
BC117670.1 | - | 1 | 2818 | 40038569 | ||
BC167155.1 | - | 2 | 2922 | 8862564 | ||
CR456387.1 | - | 1 | 2508 |