Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256702.1 | NP_001243631.1 | 14 | 1602 | ||
NM_006537.3 | NP_006528.2 | 15 | 1734 | |||
NR_046341.1 | 16 | 1887 | ||||
NR_046342.1 | 16 | 1850 | ||||
XM_017022763.1 | XP_016878252.1 | 17 | 2733 | |||
XM_017022764.1 | XP_016878253.1 | 16 | 2669 | |||
XM_017022765.1 | XP_016878254.1 | 16 | 1875 | |||
XM_017022766.1 | XP_016878255.1 | 17 | 1918 | |||
XM_017022767.1 | XP_016878256.1 | 18 | 2849 | |||
XM_017022768.1 | XP_016878257.1 | 18 | 2923 | |||
GenBank mRNA | AF073344.1 | - | 15 | 1601 | ||
AK022931.1 | - | 13 | 1320 | |||
AK301236.1 | - | 14 | 1518 | |||
AK301563.1 | - | 9 | 1033 | |||
AK307810.1 | - | 15 | 1493 | |||
AK309382.1 | - | 14 | 1555 | |||
AK314223.1 | - | 15 | 1653 | |||
AY461579.1 | - | 16 | 1763 | |||
BC018113.2 | - | 15 | 1599 | 3916811 | ||
BC065300.1 | - | 15 | 1634 | 6146202 | ||
BC107137.1 | - | 15 | 1594 | 40025968 | ||
BC107138.1 | - | 15 | 1594 | 40025969 | ||
BT007269.1 | - | 15 | 1505 | |||
BX647760.1 | - | 16 | 1628 |