Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NR1I3 SIRNA Assays ›
Gene Symbol:
NR1I3
Gene Name:
nuclear receptor subfamily 1 group I member 3
Gene Aliases:
CAR, CAR1, MB67
Chromosome Location:
Chr.1: 161229666 - 161238623 on Build GRCh38
UniGene ID:
Hs.349642
Species:
Human
siRNA ID
HSS115122
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NR1I3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077469.2
NP_001070937.1
5
688
NM_001077470.2
NP_001070938.1
4
548
NM_001077471.2
NP_001070939.1
5
688
NM_001077472.2
NP_001070940.1
4
548
NM_001077473.2
NP_001070941.1
4
548
NM_001077474.2
NP_001070942.1
5
688
NM_001077475.2
NP_001070943.1
4
548
NM_001077476.2
NP_001070944.1
4
548
NM_001077477.2
NP_001070945.1
4
548
NM_001077478.2
NP_001070946.1
5
688
NM_001077479.2
NP_001070947.1
4
548
NM_001077480.2
NP_001070948.1
5
688
NM_001077481.2
NP_001070949.1
5
688
NM_001077482.2
NP_001070950.1
5
688
NM_005122.4
NP_005113.1
5
688
XM_005245693.4
XP_005245750.1
4
961
XM_005245694.4
XP_005245751.1
4
960
XM_005245697.4
XP_005245754.1
5
571
XM_011510237.2
XP_011508539.1
4
958
GenBank mRNA
AB307702.1
-
4
485
AK303757.1
-
4
541
AK303796.1
-
5
722
AK310773.1
-
3
259
AK316208.1
-
5
680
AY572806.1
-
5
579
AY572807.1
-
5
508
AY572808.1
-
4
439
AY572809.1
-
3
269
AY572810.1
-
5
579
AY572811.1
-
4
439
AY572812.1
-
4
439
AY572813.1
-
5
579
AY572814.1
-
3
269
AY572815.1
-
3
269
AY572816.1
-
4
439
AY572817.1
-
4
439
AY572818.1
-
4
439
AY572819.1
-
5
579
AY572820.1
-
4
349
AY572826.1
-
4
409
AY572827.1
-
4
439
BC069626.1
-
5
641
7262456
BC121120.1
-
5
589
40120664
BC121121.1
-
5
589
40120665
DQ022681.1
-
4
485
HQ692838.1
-
4
485
HQ692839.1
-
4
485
HQ692840.1
-
4
485
HQ692841.1
-
4
485
HQ709178.1
-
4
485
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
transcription initiation from RNA polymerase II promoter
signal transduction
androgen receptor signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
androgen receptor activity
thyroid hormone receptor activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline