Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042388.2 | NP_001035847.1 | 11 | 1638 | ||
NM_005134.3 | NP_005125.1 | 11 | 1587 | |||
NR_052003.1 | 10 | 1480 | ||||
XM_011525775.2 | XP_011524077.1 | 11 | 1764 | |||
XM_011525776.1 | XP_011524078.1 | 11 | 1628 | |||
XM_011525779.2 | XP_011524081.1 | 8 | 1431 | |||
XM_017026105.1 | XP_016881594.1 | 10 | 1605 | |||
XM_017026106.1 | XP_016881595.1 | 10 | 1442 | |||
XM_017026107.1 | XP_016881596.1 | 10 | 1457 | |||
GenBank mRNA | AF111106.1 | - | 11 | 1557 | ||
AF200478.1 | - | 11 | 1537 | |||
AK025509.1 | - | 3 | 745 | |||
AK027064.1 | - | 11 | 1589 | |||
AK292538.1 | - | 11 | 1518 | |||
AK293869.1 | - | 10 | 1378 | |||
AK295533.1 | - | 1 | 247 | |||
AK315106.1 | - | 11 | 1545 | |||
BC060829.1 | - | 11 | 1539 | 30342594 |