Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NAALAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300930.1 | NP_001287859.1 | 13 | 1439 | ||
NM_005467.3 | NP_005458.1 | 14 | 1538 | |||
XM_017017043.1 | XP_016872532.1 | 16 | 1704 | |||
XM_017017044.1 | XP_016872533.1 | 15 | 1606 | |||
XM_017017045.1 | XP_016872534.1 | 14 | 1541 | |||
GenBank mRNA | AJ012370.1 | - | 14 | 1474 | ||
AK075390.1 | - | 14 | 1513 | |||
BC096316.3 | - | 14 | 1450 | 40007638 | ||
BC096317.3 | - | 14 | 1449 | 40007639 | ||
BC096318.3 | - | 14 | 1449 | 40007641 |