Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177465.2 | NP_001170936.1 | 3, 4 | 2168 | ||
NM_001177466.2 | NP_001170937.1 | 4, 5 | 2282 | |||
NM_005491.4 | NP_005482.2 | 5, 6 | 2357 | |||
XM_006724802.3 | XP_006724865.1 | 7, 8 | 2705 | |||
XM_006724803.3 | XP_006724866.1 | 6, 7 | 2630 | |||
XM_011531092.2 | XP_011529394.1 | 5, 6 | 2338 | |||
XM_011531093.2 | XP_011529395.1 | 6, 7 | 2468 | |||
XM_011531095.2 | XP_011529397.1 | 6, 7 | 2582 | |||
XM_017029188.1 | XP_016884677.1 | 7, 8 | 2577 | |||
XM_017029189.1 | XP_016884678.1 | 6, 7 | 2455 | |||
GenBank mRNA | AK094563.1 | - | 1 | 647 | ||
AK125184.1 | - | 1 | 1677 | |||
AK294478.1 | - | 3, 4 | 2027 | |||
AK297504.1 | - | 2, 3 | 738 | |||
AK302612.1 | - | 7, 8 | 2129 | |||
AK315217.1 | - | 5, 6 | 2230 | |||
BC136324.1 | - | 4, 5 | 2267 | 9020311 | ||
BX537560.1 | - | 1 | 4028 | |||
DA527328.1 | - | 3, 4 | 436 | |||
U46023.1 | - | 5, 6 | 2358 |