Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CST8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281730.1 | NP_001268659.1 | 2 | 374 | ||
NM_005492.3 | NP_005483.1 | 2 | 533 | |||
GenBank mRNA | AF059244.1 | - | 2 | 512 | ||
AW665128.1 | - | 3 | 387 | |||
BC069496.1 | - | 2 | 323 | 7262200 | ||
BC069536.1 | - | 2 | 323 | |||
BC105113.1 | - | 1 | 205 | 8144116 | ||
BC105119.1 | - | 1 | 205 | 8144122 | ||
BX096109.1 | - | 2 | 373 |