Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286121.1 | NP_001273050.1 | 13 | 1614 | ||
NM_005495.2 | NP_005486.1 | 12 | 1514 | |||
XM_011514211.1 | XP_011512513.1 | 12 | 1553 | |||
XM_011514213.2 | XP_011512515.1 | 11 | 1306 | |||
XM_011514214.2 | XP_011512516.1 | 10 | 1210 | |||
XM_011514217.2 | XP_011512519.1 | 8 | 1012 | |||
XM_011514218.2 | XP_011512520.1 | 8 | 842 | |||
XM_011514219.2 | XP_011512521.1 | 7 | 850 | |||
XM_017010150.1 | XP_016865639.1 | 11 | 1343 | |||
XM_017010151.1 | XP_016865640.1 | 8 | 929 | |||
GenBank mRNA | AB020527.1 | - | 12 | 1503 | ||
AK024903.1 | - | 12 | 1514 | |||
AK293354.1 | - | 7 | 784 | |||
AK308500.1 | - | 3 | 183 | |||
BC109207.1 | - | 7 | 1920 | |||
BC109208.1 | - | 12 | 1542 | 40006623 |