Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NR1H3 SIRNA Assays ›
Gene Symbol:
NR1H3
Gene Name:
nuclear receptor subfamily 1 group H member 3
Gene Aliases:
LXR-a, LXRA, RLD-1
Chromosome Location:
Chr.11: 47248300 - 47269033 on Build GRCh38
UniGene ID:
Hs.438863
Species:
Human
siRNA ID
HSS115297
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NR1H3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130101.2
NP_001123573.1
8
1217
NM_001130102.2
NP_001123574.1
8
1206
NM_001251934.1
NP_001238863.1
9
1345
NM_001251935.1
NP_001238864.1
9
1386
NM_005693.3
NP_005684.2
9
1397
XM_005252705.1
XP_005252762.1
10
1350
XM_005252706.1
XP_005252763.1
10
1467
XM_005252707.4
XP_005252764.1
9
1483
XM_005252709.1
XP_005252766.1
10
1351
XM_005252710.1
XP_005252767.1
9
1249
XM_005252713.3
XP_005252770.1
8
1092
XM_005252715.2
XP_005252772.1
8
1077
XM_005252716.3
XP_005252773.1
7
941
XM_005252718.3
XP_005252775.1
6
762
XM_006718112.1
XP_006718175.1
10
1339
XM_006718113.1
XP_006718176.1
10
1350
XM_006718115.1
XP_006718178.1
9
1357
XM_006718116.1
XP_006718179.1
9
1266
XM_011519805.2
XP_011518107.1
9
1269
XM_011519806.1
XP_011518108.1
7
1053
XM_011519807.1
XP_011518109.1
7
971
XM_011519808.2
XP_011518110.1
6
737
XM_017017058.1
XP_016872547.1
8
1182
GenBank mRNA
AB307698.1
-
8
1147
AK122661.1
-
8
2385
AK290614.1
-
9
1352
AK302132.1
-
9
1334
AK313311.1
-
9
1345
BC008819.2
-
8
1136
3957848
BC041172.1
-
8
1188
5744574
BT019411.1
-
8
1147
HQ692819.1
-
8
1147
U22662.1
-
8
1182
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
negative regulation of macrophage derived foam cell differentiation
positive regulation of triglyceride biosynthetic process
positive regulation of receptor biosynthetic process
positive regulation of cholesterol efflux
negative regulation of cholesterol storage
intracellular receptor signaling pathway
positive regulation of cellular protein metabolic process
negative regulation of lipid transport
positive regulation of cholesterol transport
response to progesterone
positive regulation of toll-like receptor 4 signaling pathway
cholesterol homeostasis
regulation of circadian rhythm
negative regulation of macrophage activation
apoptotic cell clearance
steroid hormone mediated signaling pathway
positive regulation of fatty acid biosynthetic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of pinocytosis
negative regulation of inflammatory response
positive regulation of lipoprotein lipase activity
lipid homeostasis
sterol homeostasis
negative regulation of interferon-gamma-mediated signaling pathway
triglyceride homeostasis
cellular response to lipopolysaccharide
negative regulation of pancreatic juice secretion
negative regulation of secretion of lysosomal enzymes
regulation of cholesterol homeostasis
positive regulation of cholesterol homeostasis
DNA binding
steroid hormone receptor activity
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
zinc ion binding
cholesterol binding
sterol response element binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0