Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005723.3 | NP_005714.2 | 1 | 470 | ||
XM_005262680.1 | XP_005262737.1 | 1 | 470 | |||
GenBank mRNA | AF065389.1 | - | 1 | 382 | ||
AK315723.1 | - | 1 | 437 | |||
BC009704.1 | - | 1 | 368 | 3895933 | ||
CR541809.1 | - | 1 | 35 | |||
CR541829.1 | - | 1 | 35 |